Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Arsenite, 1% (w/v), Ricca Chemical
SDP

Catalog No. 7140132 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71401-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71401-32 Supplier Ricca Chemical Company Supplier No. 7140.132
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7784-46-5
Molecular Formula AsNaO2
Molecular Weight (g/mol) 129.909
InChI Key PTLRDCMBXHILCL-UHFFFAOYSA-M
Synonym sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c
PubChem CID 443495
ChEBI CHEBI:29678
IUPAC Name sodium;oxoarsinite
SMILES [O-][As]=O.[Na+]

Specifications

Color Colorless
CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Physical Form Liquid
Quantity 1 L
Percent Purity 1% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Arsenite
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.