Learn More
Selleck Chemical LLC isoleucine S3752-25mg
SDP
Supplier:  Selleck Chemical LLC S375225MG
Encompass_Preferred
Isoleucine (Ile) is a non-polar uncharged (at physiological pH) aliphatic amino acid that is used in the biosynthesis of proteins
Catalog No. 50-310-8374
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.