Learn More
Description
- Product is for educational use
- Available in 500 g, 2.5 kg, and 3 kg
This product is for educational use - no C of A is available.
Specifications
Specifications
| Quantity | 500 g |
| Assay Percent Range | 99% w/v |
| Linear Formula | NaHCO3 |
| Formula Weight | 84.01 |
| Packaging | Lab Grade |
| Chemical Name or Material | Sodium Bicarbonate |
Safety and Handling
| ShelfLife | 5 years |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
