Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bicarbonate, 99.5%, MP Biomedicals™

Catalog No. ICN19549701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19549701 1 kg
MP21954975 500 g
ICN19549703 3 kg
ICN19549705 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19549701 Supplier MP Biomedicals, Inc Supplier No. 0219549701
Add to Cart
Add to Cart

Sodium bicarbonate ≥99.5%, White Powder

  • It is broadly used as a physiological buffer in cell culture applications.
  • The pKa for bicarbonate <-> carbonic acid reaction is 6.4 The pKa for bicarbonate <-> carbonate reaction is 10.3.
  • It has two pKas, 6.4 and 10.3. The compound can react with both acids and bases, therefore it can be used as a neutralizer.
  • It may be used to remove or wash acidic impurities from a crude liquid to yield a purer sample.
  • it reacts with the carboxyl groups to give a fast-forming and effervescent CO2 formation which has been used to test for the presence of carboxylic groups in proteins.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.5%
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
Formula Weight 84.01
Percent Purity 99.5%
Density 2.159g/mL
Chemical Name or Material Sodium bicarbonate
EINECSNumber 205-633-8
Recommended Storage Store at room temperature.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.