Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate, Powder, BiotechGrade, 99.7-100.3%, Spectrum™ Chemical
SDP

Catalog No. 18606324 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-324 6 x 500 g Poly Bottle
18-606-323 500 g Poly Bottle
18-606-325 2.5 kg Poly Bottle
18-606-326 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-324 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1147500GMCS6
Add to Cart
Add to Cart

S1147, 144-55-8, NaHCO3

Spectrum™ Chemical Sodium Bicarbonate, Powder, BiotechGrade is a commonly used laboratory chemical that can react with both acids and bases, therefore it can be used as a neutralizer. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

CAS Max % 1
Quantity 6 x 500 g
Percent Purity 99.7 to 100.3%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.