Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate, Powder, USP, EP, BP, JP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606338 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
25 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-338 45 kg Fiber Drum
18-606-335 2.5 kg Poly Bottle
18-606-334 6 x 500 g Poly Bottle
18-606-336 4 x 2.5 kg Poly Bottle
18-606-337 12 kg Poly Pail
18-606-339 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-606-338 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO12545KGBL
Add to Cart
Add to Cart

SO125, 144-55-8, NaHCO3

Spectrum™ Chemical Sodium Bicarbonate, Powder, USP, EP, BP, JP is used as an antacid to treat heartburn, indigestion, and upset stomach. All Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

pH 7.9 to 8.4
CAS Max % 1
Quantity 45 kg
Percent Purity 99 to 100.5%
Grade Multi-Compendial
Loss on Drying 0.0025
Packaging Fiber Drum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.