Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate, Purified (Sodium Hydrogen Carbonate), Reagents

Catalog No. 01337059
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-059 50 lb.
01-337-062 12 kg
01-337-061 2.5 kg
01-337-060 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-059 Supplier Reagents Holdings Llc Supplier No. C230850050F5
Add to Cart
Add to Cart

Sodium Bicarbonate, Purified (Sodium Hydrogen Carbonate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym Sodium hydrogen carbonate
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270°C
Color Colorless/White
Physical Form Solid
Quantity 50 lb.
Grade Pure
Packaging Fiber Drum
Specific Gravity 2.159
Chemical Name or Material Sodium Bicarbonate, Purified (Sodium Hydrogen Carbonate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.