Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bisulfite, ACS Reagent , Reagents

Catalog No. 01337755
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-755 500 g
01-337-757 12 kg
01-337-756 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-755 Supplier Reagents Holdings Llc Supplier No. C1310000500C2
Add to Cart
Add to Cart

Sodium Bisulfite, ACS Reagent , ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7631-90-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
Synonym Sodium Hydrogen Sulfite
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

Boiling Point 146°C
Melting Point 5°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.48
Chemical Name or Material Sodium Bisulfite, ACS Reagent *This reagent is usually a mixture containing some Sodium Metabisulfite (CAS# 7681-57-4).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.