Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borohydride, ≥99%, Solstice

Catalog No. 60047349 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-349 100 g Glass bottle
60-047-350 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-349 Supplier Solstice Supplier No. 71321100G
Add to Cart
Add to Cart

TraceSELECT™, for the determination of hydride formers by AAS,≥99%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Boiling Point >400°C
Melting Point >360°C
Color White
Physical Form Prill or Powder
Quantity 100 g
Linear Formula NaBH4
UN Number UN1426
Formula Weight 37.83g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Glass bottle
Vapor Pressure >0.000054 Pa
Density 1.074g/cm3
Chemical Name or Material Sodium borohydride
Show More Show Less
Hazard Statement P201-P231 + P232-P280-P308 + P313-P370 + P378-P402 + P404
DOTInformation 4.3 / PGI
RTECSNumber 241-004-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.