Sodium bromide, 99.9955% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1086014

Quantity 25g

 View more versions of this product

Catalog No. AA1086014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7647-15-6
    Physical Form Powder
    Assay Percent Range 99.9955% (metals basis)
    Molecular Formula NaBr
    Formula Weight 102.89
    MDL Number MFCD00003475
    Chemical Name or Material Sodium bromide

    Specifications

    Formula Weight 102.89
    Quantity 25g
    Color White
    Density 3.203g/cm3 at 20°C
    EINECS Number 231-599-9
    Physical Form Powder
    Molecular Formula NaBr
    MDL Number MFCD00003475
    CAS 7647-15-6
    Assay Percent Range 99.9955% (metals basis)
    Appearance Powder/flake/crystalline/beads
    Chemical Name or Material Sodium bromide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit