Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bromide, ACS reagent

Catalog No. AC212670025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle

CAS: 7647-15-6 | BrNa | 102.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390.0°C
Melting Point 755.0°C
Quantity 2.5 kg
Linear Formula NaBr
Fieser 12,445; 13,46
Merck Index 15,8729
Solubility Information Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C), 1210g/L water (100°C), 1g/6 mL methanol, 7g/100 mL 95% ethanol
IUPAC Name sodium;bromide
Formula Weight 102.89
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium bromide
Show More Show Less
EINECSNumber 231-599-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.