Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bromide, ACS Reagent, Reagents

Catalog No. 01337749
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-749 2.5 kg
01-337-748 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-749 Supplier Reagents Holdings Llc Supplier No. C13110002.5A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Bromide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
IUPAC Name sodium bromide
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390°C
Melting Point 755°C
Color Colorless/White
Physical Form Solid
Quantity 2.5 kg
Percent Purity ≥99.99%
Grade ACS
Packaging Poly Bottle
Specific Gravity 3.21
Chemical Name or Material Sodium Bromide, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.