Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium cacodylate, 0.1M buffer soln., pH 6.8, Thermo Scientific Chemicals

Catalog No. AAJ62202AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62202AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62202AE Supplier Thermo Scientific Chemicals Supplier No. J62202AE
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6131-99-3 | Molecular Weight: 214.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-99-3
Molecular Weight (g/mol) 214.024

Specifications

Concentration 0.1M buffer soln.
pH 6.8
Physical Form Liquid
Quantity 100 mL
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.