Learn More
TARGETMOL CHEMICALS INC SB 242235 10MG
Supplier: TARGETMOL CHEMICALS INC T407810MG
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. SB 242235 is a potent and selective p38 MAP kinase inhibitor that may be an effective therapy for cytokine-mediated diseases. Purity 99.68%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.