Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, 0.0500 N (N/20), 1 mL = 2.5 mg CaCO3, Ricca Chemical
SDP

Catalog No. 718532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7185-32 1 L Natural Poly Bottle
7185-16 500 mL Natural Poly Bottle
R71850004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 7185-32 Supplier Ricca Chemical Company Supplier No. 718532
Add to Cart
Add to Cart

APHA/EPA for Alkalinity Testing

Specifications

CAS 7732-18-5
Grade Laboratory
Packaging Natural Poly Bottle
Methods APHA/EPA for Alkalinity
pH 11 to 11.5
Quantity 1 L
Chemical Name or Material Sodium Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.