Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, Anhydrous, Purified (Soda Ash), Reagents

Catalog No. 01337041
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
50 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-041 50 lb.
01-337-044 10 kg
01-337-043 2.5 kg
01-337-042 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-041 Supplier Reagents Holdings Llc Supplier No. C231210050F5
Add to Cart
Add to Cart

Sodium Carbonate, Anhydrous, Purified (Soda Ash), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym Soda ash, Disodium carbonate
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Melting Point 851°C
Color Colorless/White
Physical Form Solid
Quantity 50 lb.
Grade Pure
Packaging Fiber Drum
Specific Gravity 2.53
Chemical Name or Material Sodium Carbonate, Anhydrous, Purified (Soda Ash)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.