Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate decahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., E500, 99.5-100.5% (ex dried subst.), Solstice

Catalog No. 6001980 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-80 2.5 kg
60-019-79 1 kg
60-044-794 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-019-80 Supplier Solstice Supplier No. 134142.5KG
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, FCC, Ph. Eur., E500, 99.5-100.5% (ex dried subst.)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-1-6132
Molecular Formula CH20Na2O13
Molecular Weight (g/mol) 286.14
MDL Number MFCD00149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

Quantity 2.5 kg
Linear Formula Na2CO3 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 286.14g/mol
Percent Purity 99.5 to 100.5% (ex dried subst.)
Grade puriss.
Chemical Name or Material Sodium Carbonate Decahydrate, Meets analytical specification of Ph. Eur.BP, FCC
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 207-838-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.