Learn More
Sodium carbonate, Powder, ≥99.5%, ACS reagent, Solstice
Shop All Honeywell Research Chemicals ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Melting Point | 851°C (literature) |
| CAS Min % | ≥90.0000% |
| CAS Max % | ≤100.0000% |
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Linear Formula | Na2CO3 |
| Beilstein | 4154566 |
| IUPAC Name | disodium carbonate |
| Formula Weight | 105.99 |
| Percent Purity | >99.5% |
| Show More |
Safety and Handling
| ShelfLife | 1260 days from date of manufacture |
| EINECSNumber | 207-838-8 |
| Recommended Storage | Room Temp |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.