Learn More
Sodium carbonate/Sodium hydroxide, IC eluent, Concentrate (20x) for Metrosep A Supp 16, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 497-19-8 |
| Quantity | 2.5 L |
| Concentration or Composition (by Analyte or Components) | 150 mM Na2 CO3 aq. soln. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.