Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chlorate, 99+%, extra pure

Catalog No. AC223220010
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223220010 1 kg Plastic bottle
AC223222500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223220010 Supplier Thermo Scientific Chemicals Supplier No. 223220010
Add to Cart

CAS: 7775-09-9 | ClNaO3 | 106.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-09-9
Molecular Formula ClNaO3
Molecular Weight (g/mol) 106.44
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
Synonym sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
PubChem CID 516902
ChEBI CHEBI:65242
SMILES [Na+].[O-][Cl](=O)=O

Specifications

Melting Point 248.0°C to 261.0°C
Color White
Physical Form Crystals
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula NaClO3
Fieser 01,1056
Merck Index 15,8733
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and glycerin, 7.7g/L ethanol (20°C)
IUPAC Name sodium chlorate
Formula Weight 106.44
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 2.49
Density 2.4900g/mL
Chemical Name or Material Sodium chlorate
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
May cause fire or explosion; strong oxidizer.
missing translation for 'healthHazard3' GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 231-887-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.