Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, 0.9% w/w, Reagents

Catalog No. 01333664
Change view
Click to view available options
Quantity:
10 L
20 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-664 20 L
01-333-666 10 L
01-333-665 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-333-664 Supplier Reagents Holdings Llc Supplier No. CS14020020F
Add to Cart
Add to Cart

Sodium Chloride, 0.9% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-14-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Table salt solution
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH 5 to 9
Physical Form Liquid
Quantity 20 L
Percent Purity 0.9%
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.0094
Chemical Name or Material Sodium Chloride, 0.9% w/w
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.