Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, 9% (w/v), Ricca Chemical
SDP

Catalog No. 721591 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7215-91 4 L Natural Poly Bottle
7215-932 1 L Natural Poly Bottle
7215-92-5 10 L Cubitainer
7215-95 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 7215-91 Supplier Ricca Chemical Company Supplier No. 7215.91
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Colorless
CAS Min % 90.0, 8.1
CAS Max % 94.0, 8.43
Physical Form Liquid
Quantity 4 L
Percent Purity 9% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 9% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Chloride
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.