Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, 99.0%, MilliporeSigma™

Catalog No. 50947346 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
159 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-346 159 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-346 Supplier MilliporeSigma Supplier No. SX042020

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C (1013 hPa)
Melting Point 801°C
pH 4.5 to 7.0 (100 g/L, H2O, 20°C)
Quantity 159 kg
Assay Percent Range 99.0% min
Solubility Information 358 g/L
Packaging Fiber Drum
Vapor Pressure 1.3 hPa (865°C)
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.