Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride ACS AR Crystal, Macron Fine Chemicals™

Catalog No. MK7581212 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-7581-212 2.5 kg Poly
MK-7581-500 500 g POLYSTORMOR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-7581-212 Supplier Avantor Supplier No. 758106
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Physical Form Crystalline
Quantity 2.5 kg
Formula Weight 58.44
Grade ACS
Packaging Poly
Chemical Name or Material Sodium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.