Learn More
Specifications
Specifications
| Quantity | 500 g |
| Linear Formula | NaCl |
| Formula Weight | 58.44 |
| Grade | ACS |
| Packaging | Poly bottle |
| Vapor Pressure | 1.3 hPa (865°C) |
| Density | 2.17g/cm3 (20°C); Bulk Density: 1140 kg/m3 |
| Chemical Name or Material | Sodium Chloride, Certipure™, Secondary Reference Material For Argentometry |
Safety and Handling
| Recommended Storage | Store at +15°C to +25°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.