Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCS043050F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS043050F 50 kg
RDCS0430500 500 g
RDCS043025A 2.5 kg
RDCS043012F 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RDCS043050F Supplier Ricca Chemical Company Supplier No. RDCS043050F4
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride

Specifications

pH 5.0-9.0
CAS Max % 100
Physical Form Adhering Powder and/or Lumps
Quantity 50 kg
SMILES [Na+].[Cl-]
Percent Purity min 99%
Grade ACS
Packaging Fiber Drum
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.