Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Cell Culture Reagent Grade, MP Biomedicals™

Catalog No. MP219473810 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219473810 10 kg
MP21947385 500 g
MP219473801 1 kg
MP219473805 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP219473810 Supplier MP Biomedicals, Inc Supplier No. 0219473810
Add to Cart
Add to Cart

Used in a wide variety of biochemical applications. Sodium Chloride, Cell Culture Reagent Grade, MP Biomedicals is a chemical found widely in nature. It is considered an essential nutrient. This product is used as a diluent, in protein crystallization, in

Sodium chloride is used in a wide variety of biochemical applications, including:

  • In protein crystallization
  • Intravenous fluids (normal saline is 0.9% w/v in water10)
  • Density gradients
  • A diluent to increase ionic strength in buffers or culture media
  • In salt-out procedures in the isolation of DNA
  • In high concentrations for preservation of foods, since bacteria cannot grow in high salt conditions

A salt-ice mixture in the ratio of approximately 33g sodium chloride to 100g ice (at −1°C) will drop in temperature to as low as &minus’21°C, depending on the rate of stirring and the size of the ice chunks. Excess amounts of sodium chloride can destroy electrolyte balance and cause death in most animals, including humans.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Physical Form Powder
Quantity 10 kg
Solubility Information Soluble in water (357mg/mL at 25°C; 384mg/mL at 100°C), glycerol (100mg/mL); very slightly soluble in ethanol. Almost insoluble in concentrated HCl.
IUPAC Name sodium chloride
Formula Weight 58.44g/mol
Percent Purity ≥99.5%
Grade Cell Culture Reagent
Recommended Storage Store at Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.