Learn More
Sodium Chloride (colorless to white crystalline granules/USP/FCC/EP/BP/JP), Fisher Chemical™
Shop All Fisherbrand Products

Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Boiling Point | 1461°C |
| Melting Point | 801°C |
| Color | White |
| pH | 5.0 to 8.0 |
| Physical Form | Solid |
| Quantity | 12 kg |
| Formula Weight | 58.44 |
| Percent Purity | 99.0 to 100.5% (EP) |
| Grade | BP/EP/FCC/JP/USP |
| Identification | Pass Test (BP, SP, FCC, JP, USP), Pass Test (BP, SP, FCC, Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (USP) |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention. NFPA Health:1 Flammability:0 Instability:1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.