Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride (colorless to white crystalline granules/USP/FCC/EP/BP/JP), Fisher Chemical™
GSA_VA

Catalog No. S642500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
350 lb.
Packaging:
Poly Bottle
Poly Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S642-500 500 g Poly Bottle
S642-212 2.5 kg Poly Bottle
S642-12 12 kg Poly Pail
S642-350LB 350 lb. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. S642-500
Add to Cart
This item is not returnable. View return policy

This product is not intended for dialysis, parenteral or injectable use without further processing

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 500 g
Formula Weight 58.44
Percent Purity 99.0 to 100.5% (BP)
Grade BP/EP/FCC/JP/USP
Identification Pass Test (BP, SP, FCC, JP, USP), Pass Test (BP, SP, FCC, Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (USP)
Loss on Drying ≤0.5% (at 110°C, FCC), ≤0.5% (BP/BP, EP/EP, JP/JP, USP/USP), 0.5% max. (BP), 0.5% max. (EP), 0.5% max. (FCC), 0.5% max. (JP), 0.5% max. (USP)
Packaging Poly Bottle
Vapor Pressure 1mmHg at 440°C
Density 2.165g/cm³
Chemical Name or Material Sodium Chloride
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.