Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Granular, BiotechGrade, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18606405 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-405 500 g Poly Bottle
18-606-407 2.5 kg Poly Bottle
18-606-406 6 x 500 g Poly Bottle
18-606-408 4 x 2.5 kg Poly Bottle
18-606-409 12 kg Poly Pail
18-606-410 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-606-405 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1249500GM
Add to Cart
Add to Cart

S1249, 7647-14-5, NaCl

Spectrum™ Chemical Sodium Chloride, Granular, BiotechGrade is an ionic compound that is also known as salt. It is responsible for the salinity level of the ocean. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99 to 101%
Grade Biotech
Loss on Drying 0.005
Odor Odorless
Packaging Poly Bottle
Appearance White, crystallin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.