Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Granular, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606412 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
250 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-412 6 x 500 g Poly Bottle
18-606-414 4 x 2.5 kg Poly Bottle
18-606-415 12 kg Poly Pail
18-606-416 45 kg Fiber Drum
18-606-417 250 lb. Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-606-412 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO155500GMCS6

SO155, 7647-14-5, NaCl

Spectrum™ Chemical Sodium Chloride, Granular, USP is used in the formulation of oral hygiene products, shampoos, fragrance, skin, hair, nail, cleansing, suntan, makeup and bath products. This grade is specifically used in oral care applications. This product is not sterile and not intended for use in the manufacture of injectable dosage forms, peritoneal dialysis solutions, hemodialysis solutions or hemofiltration solutions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

CAS Max % 1
Quantity 6 x 500 g
Percent Purity 99 to 100.5%
Loss on Drying 0.005
Packaging Poly Bottle
Appearance Clear
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.