Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chloride, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al, Solstice

Catalog No. 6001985 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-85 1 kg
60-019-87 5 kg
60-044-798 6 x 1 kg
60-044-797 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-019-85 Supplier Solstice Supplier No. 134231KG
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Quantity 1 kg
Linear Formula NaCl
UN Number NONH for all modes of transport
Beilstein 3534976
IUPAC Name sodium chloride
Formula Weight 58.44g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Sodium Chloride, Meets analytical specification of Ph. Eur.,BP, USP
EINECSNumber 231-598-3
RTECSNumber VZ4725000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.