Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Cholate, Reagent, 95%, Spectrum™ Chemical
SDP

Catalog No. 18606440 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-440 500 g Poly Bottle
18-606-437 5 g Amber Glass Bottle
18-606-438 25 g Poly Bottle
18-606-439 100 g Poly Bottle
18-606-441 1 kg Poly Pail
18-606-442 5 kg Poly Pail
18-606-443 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-606-440 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO162500GM
Add to Cart
Add to Cart

SO162, 361-09-1, C24H39NaO5

Spectrum™ Chemical Sodium Cholate, Reagent The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 361-09-1
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.56
MDL Number MFCD00150749,MFCD00064138
InChI Key NRHMKIHPTBHXPF-TUJRSCDTSA-M
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
SMILES [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 95%
Grade Reagent
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.