Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chromate, 2% (w/v), Ricca Chemical
SDP

Catalog No. 723432 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
723432 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 723432 Supplier Ricca Chemical Company Supplier No. 723432
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10034-82-9 , 7732-18-5
Molecular Formula CrH8Na2O8
Molecular Weight (g/mol) 234.032
InChI Key AVHUVTPOXZEIRG-UHFFFAOYSA-N
Synonym sodium chromate tetrahydrate, cro4.2na.4h2o, acmc-20ajol, disodium chromate tetrahydrate, ksc492q3d, disodium chromate 2-tetrahydrate, sodium chromate tetrahydrate, reagent grade, sodium chromate tetrahydrate, p.a
PubChem CID 22146515
IUPAC Name disodium;dioxido(dioxo)chromium;tetrahydrate
SMILES O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Yellow
CAS Min % 96.0, 1.92
CAS Max % 100.0, 2.0
Percent Purity 2%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH Alkaline
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 2% (w/v)
Chemical Name or Material Sodium Chromate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.