accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOCrOOO

CAS RN 10034-82-9

IUPAC Name: disodium dioxochromiumbis(olate)
Synonyms: sodium chromate
Molecular Weight (g/mol): 161.97
Molecular Formula: CrNa2O4
InChi Key: PXLIDIMHPNPGMH-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[O-][Cr]([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
Grade 
  • (1)
  • (4)
  • (7)
  • (2)
Physical Form 
  • (2)
  • (4)
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (9)
  • (1)
  • (2)
  • (9)
  • (4)
  • (2)
  • (5)
  • (9)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (7)
  • (2)
  • (2)
  • (4)
  • (2)
  • (5)
  • (2)
  • (1)
  • (5)
  • (1)