Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chromate Tetrahydrate, Crystal, 99%, Spectrum™ Chemical
SDP

Catalog No. 18613442 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-442 12 kg Poly Bottle
18-613-438 25 g Poly Bottle
18-613-439 125 g Poly Bottle
18-613-440 500 g Poly Bottle
18-613-441 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-613-442 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S124512KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10034-82-9

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 12 kg
Molecular Weight (g/mol) 234.06
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Chromate Tetrahydrate, Crystal
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.