Learn More
Boston Bioproducts Inc Citrate Buffer (0.5 M, pH 6.0) - 250ml
Supplier:  Boston Bioproducts Inc BB90250ML
Encompass_Preferred
The Citrate Buffer comprising of Sodium citrate dihydrate and Citric acid is commonly used to maintain pH levels within the range of 3.0 to 6.2. This buffer supports several applications in molecular biology, pathology, histology, immunohistochemistry, and RNA isolation. It also aids in the detection of antigens in fixed tissue preparations as it induces the breaking cross-links between antigens and components in the fixation medium.
Catalog No. NC1711672
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.