Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sodium citrate, 0.5M buffer soln., pH 6.5

Catalog No. AAJ63888AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63888AK 250 mL
AAJ63888AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63888AK Supplier Thermo Scientific Chemicals Supplier No. J63888AK
Add to Cart

Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 

Solubility
Soluble in water.

Notes
20X SSC Solution (RNase-free) should be stored at room temperature. This reagent should remain stable for at least 1 year in its unopened container.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium citrate buffer soln.
Color Colorless
Concentration 0.5 M
pH 6.5
Physical Form Liquid
Quantity 250 mL
Odor Odorless
MDL Number MFCD00012462
Solubility Information Soluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.