Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate, Dihydrate, Crystal, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18613454 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-454 500 g Poly Bottle
18-613-455 2.5 kg Poly Bottle
18-613-456 12 kg Poly Pail
18-613-457 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-613-454 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1250500GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4-3-6132

Specifications

pH 7.0 to 9.0
CAS Max % 1
Quantity 500 g
Percent Purity 99%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.