Learn More
TARGETMOL CHEMICALS INC SODIUM DEMETHYLCANTHARID 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T273650MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg and bulk. Please contact Fisher for quotes. Sodium Demethylcantharidate (Sodium norcantharidin) is extracted from Mylabris phqlarata pallas and has potent antitumor activity. purity: 99%
Catalog No. 50-257-7053