Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium deoxycholate monohydrate, 97%, Thermo Scientific Chemicals

Catalog No. AAJ6228830
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6228830 250 g
AAJ6228814 25 g
AAJ6228822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6228830 Supplier Thermo Scientific Chemicals Supplier No. J6228830
Add to Cart

Sodium deoxycholate is a bile acid salt that contains a deoxycholate group and shows choleretic activity. It is used as a biological detergent to lyse cells and to solubilize cellular and membrane components.

Sodium deoxycholate is used as a choleretic and an anionic detergent. It is useful for the extraction of membrane receptors, plasma membrane proteins, nuclei isolation and solubilizes fats as well as membrane components. It acts as an intermediate for the production of corticosteroids and microbiological diagnostic media. In addition, it acts as a catalyst in the preparation of protein extracts for immunoblotting and immunoprecipitation of radiolabeled proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Sodium deoxycholate is a bile salt and an ionic detergent
• It is a bile acid formed by bacterial action from cholate
• Sodium deoxycholate is considered to be a cholagogue and choleretic since it promotes the flow of bile into the duodenum (cholagogues) or stimulates bile production by the liver (choleretic)

Application

• In chemical approaches, sodium deoxycholate acts as an intermediate for corticosteroid production
• This compound is used to lyse cells and to extract cellular and membrane components, including membrane receptors, plasma membrane proteins, and nuclei, and allows the solubilization of fats for intestinal absorption and membrane components
• Sodium deoxycholate is also used to prepare microbiological diagnostic media and acts as a catalyst in the preparation of protein extracts for immunoblotting and immunoprecipitation of radiolabeled proteins
• In biological experiments, sodium deoxycholate has been used in the lysis buffer prepared for brown adipose tissue
• It is also used in the decellularization of aortic valvular conduit

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-92-6
Molecular Formula C24H41NaO5
Molecular Weight (g/mol) 432.58
MDL Number MFCD00150750 MFCD00064139
InChI Key NDVHNZISGVSFMP-WTCAICSISA-M
Synonym 7-Deoxycholic acid sodium salt
PubChem CID 91810855
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate hydrate
SMILES O.[Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 250 g
Beilstein 3581950
Solubility Information Soluble in waterSoluble in water,dimethyl sulfoxide and methanol; Slightly soluble in ethanol.
Formula Weight 414.6
Percent Purity 97%
Chemical Name or Material Sodium deoxycholate
Hazard Category H302-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 206-132-7
RTECSNumber FZ2250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.