Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium diphenylamine-4-sulfonate, ACS Reagent, Solstice

Catalog No. 60046211 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-211 10 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-046-211 Supplier Solstice Supplier No. 3315010G

missing translation for 'validMainframeMsg'

Indicator for the determination of the redox potential, ACS Reagent, E0 in sulfuric acid 1 mol/l = +0,83 volt, rH = 27−29

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6152-67-6
Molecular Formula C12H11NNaO3S
Molecular Weight (g/mol) 272.27
MDL Number MFCD00036468
InChI Key VMVSDVIYIMNZBS-UHFFFAOYSA-N
Synonym sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
PubChem CID 517002
IUPAC Name sodium;4-anilinobenzenesulfonate
SMILES [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Color Beige to White
Quantity 10 g
Linear Formula C12H10NNaO3S
UN Number NONH for all modes of transport
Formula Weight 271.27g/mol
Packaging Glass Bottle
Appearance Powder
Chemical Name or Material Sodium diphenylamine-4-sulfonate
RTECSNumber 228-165-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.