CAS RN 6152-67-6
Sodium diphenylamine-4-sulfonate, 98%
CAS: 6152-67-6 Molecular Formula: C12H11NNaO3S Molecular Weight (g/mol): 272.27 MDL Number: MFCD00036468 InChI Key: VMVSDVIYIMNZBS-UHFFFAOYSA-N Synonym: sodium 4-phenylamino benzenesulfonate,sodium diphenylamine-4-sulfonate,benzenesulfonic acid, 4-phenylamino-, monosodium salt,unii-3e2ux1382o,sodium 4-anilinobenzenesulfonate,diphenylamine-4-sulfonic acid sodium salt,sodium diphenylamine-p-sulfonate,n-phenylsulfanilic acid, sodium salt,4-phenylamino benzenesulfonic acid sodium salt,diphenylaminesulfonic acid sodium salt PubChem CID: 517002 IUPAC Name: sodium;4-anilinobenzenesulfonate SMILES: [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Diphenylamine-4-sulfonic acid, sodium salt, ACS reagent
CAS: 6152-67-6 Molecular Formula: C12H11NNaO3S Molecular Weight (g/mol): 272.27 MDL Number: MFCD00036468 InChI Key: VMVSDVIYIMNZBS-UHFFFAOYSA-N Synonym: sodium 4-phenylamino benzenesulfonate,sodium diphenylamine-4-sulfonate,benzenesulfonic acid, 4-phenylamino-, monosodium salt,unii-3e2ux1382o,sodium 4-anilinobenzenesulfonate,diphenylamine-4-sulfonic acid sodium salt,sodium diphenylamine-p-sulfonate,n-phenylsulfanilic acid, sodium salt,4-phenylamino benzenesulfonic acid sodium salt,diphenylaminesulfonic acid sodium salt PubChem CID: 517002 IUPAC Name: sodium;4-anilinobenzenesulfonate SMILES: [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Diphenylaminesulfonic Acid Sodium Salt, For ACS analysis, MP Biomedicals™
CAS: 6152-67-6 Molecular Formula: C12H11NNaO3S Molecular Weight (g/mol): 272.27 MDL Number: MFCD00036468 InChI Key: VMVSDVIYIMNZBS-UHFFFAOYSA-N Synonym: sodium 4-phenylamino benzenesulfonate,sodium diphenylamine-4-sulfonate,benzenesulfonic acid, 4-phenylamino-, monosodium salt,unii-3e2ux1382o,sodium 4-anilinobenzenesulfonate,diphenylamine-4-sulfonic acid sodium salt,sodium diphenylamine-p-sulfonate,n-phenylsulfanilic acid, sodium salt,4-phenylamino benzenesulfonic acid sodium salt,diphenylaminesulfonic acid sodium salt PubChem CID: 517002 IUPAC Name: 4-(phenylamino)benzene-1-sulfonic acid sodium SMILES: [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Sodium diphenylamine-4-sulfonate, ACS
CAS: 6152-67-6 Molecular Formula: C12H11NNaO3S Molecular Weight (g/mol): 272.27 MDL Number: MFCD00036468 InChI Key: VMVSDVIYIMNZBS-UHFFFAOYSA-N Synonym: sodium 4-phenylamino benzenesulfonate,sodium diphenylamine-4-sulfonate,benzenesulfonic acid, 4-phenylamino-, monosodium salt,unii-3e2ux1382o,sodium 4-anilinobenzenesulfonate,diphenylamine-4-sulfonic acid sodium salt,sodium diphenylamine-p-sulfonate,n-phenylsulfanilic acid, sodium salt,4-phenylamino benzenesulfonic acid sodium salt,diphenylaminesulfonic acid sodium salt PubChem CID: 517002 IUPAC Name: sodium;4-anilinobenzenesulfonate SMILES: [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Thermo Scientific Chemicals Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
CAS: 6152-67-6 Molecular Formula: C12H11NNaO3S Molecular Weight (g/mol): 272.27 MDL Number: MFCD00036468 InChI Key: VMVSDVIYIMNZBS-UHFFFAOYSA-N Synonym: sodium 4-phenylamino benzenesulfonate,sodium diphenylamine-4-sulfonate,benzenesulfonic acid, 4-phenylamino-, monosodium salt,unii-3e2ux1382o,sodium 4-anilinobenzenesulfonate,diphenylamine-4-sulfonic acid sodium salt,sodium diphenylamine-p-sulfonate,n-phenylsulfanilic acid, sodium salt,4-phenylamino benzenesulfonic acid sodium salt,diphenylaminesulfonic acid sodium salt PubChem CID: 517002 SMILES: [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Diphenylamine, 0.3% (w/v), Ricca Chemical
CAS: 6152-67-6 Molecular Formula: C12H11NNaO3S Molecular Weight (g/mol): 272.27 MDL Number: MFCD00036468 InChI Key: VMVSDVIYIMNZBS-UHFFFAOYSA-N Synonym: sodium 4-phenylamino benzenesulfonate,sodium diphenylamine-4-sulfonate,benzenesulfonic acid, 4-phenylamino-, monosodium salt,unii-3e2ux1382o,sodium 4-anilinobenzenesulfonate,diphenylamine-4-sulfonic acid sodium salt,sodium diphenylamine-p-sulfonate,n-phenylsulfanilic acid, sodium salt,4-phenylamino benzenesulfonic acid sodium salt,diphenylaminesulfonic acid sodium salt PubChem CID: 517002 IUPAC Name: 4-(phenylamino)benzene-1-sulfonic acid sodium SMILES: [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Sodium Diphenylamine-4-sulfonate 97.0+%, TCI America™
CAS: 6152-67-6 Molecular Formula: C12H11NNaO3S Molecular Weight (g/mol): 272.27 MDL Number: MFCD00036468 InChI Key: VMVSDVIYIMNZBS-UHFFFAOYSA-N Synonym: sodium 4-phenylamino benzenesulfonate,sodium diphenylamine-4-sulfonate,benzenesulfonic acid, 4-phenylamino-, monosodium salt,unii-3e2ux1382o,sodium 4-anilinobenzenesulfonate,diphenylamine-4-sulfonic acid sodium salt,sodium diphenylamine-p-sulfonate,n-phenylsulfanilic acid, sodium salt,4-phenylamino benzenesulfonic acid sodium salt,diphenylaminesulfonic acid sodium salt PubChem CID: 517002 IUPAC Name: 4-(phenylamino)benzene-1-sulfonic acid sodium SMILES: [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Diphenylaminesulfonic Acid Sodium Salt (Crystalline/Certified ACS), Fisher Chemical
CAS: 6152-67-6 Molecular Formula: C12H11NNaO3S Molecular Weight (g/mol): 272.27 MDL Number: MFCD00036468 InChI Key: VMVSDVIYIMNZBS-UHFFFAOYSA-N Synonym: sodium 4-phenylamino benzenesulfonate,sodium diphenylamine-4-sulfonate,benzenesulfonic acid, 4-phenylamino-, monosodium salt,unii-3e2ux1382o,sodium 4-anilinobenzenesulfonate,diphenylamine-4-sulfonic acid sodium salt,sodium diphenylamine-p-sulfonate,n-phenylsulfanilic acid, sodium salt,4-phenylamino benzenesulfonic acid sodium salt,diphenylaminesulfonic acid sodium salt PubChem CID: 517002 IUPAC Name: 4-(phenylamino)benzene-1-sulfonic acid sodium SMILES: [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
MP Biomedicals, Inc p-Diphenylaminesulfonic Acid, Na 50%,MP Biomedicals
CAS: 6152-67-6 Molecular Formula: C12H11NNaO3S Molecular Weight (g/mol): 272.27 MDL Number: MFCD00036468 InChI Key: VMVSDVIYIMNZBS-UHFFFAOYSA-N Synonym: sodium 4-phenylamino benzenesulfonate,sodium diphenylamine-4-sulfonate,benzenesulfonic acid, 4-phenylamino-, monosodium salt,unii-3e2ux1382o,sodium 4-anilinobenzenesulfonate,diphenylamine-4-sulfonic acid sodium salt,sodium diphenylamine-p-sulfonate,n-phenylsulfanilic acid, sodium salt,4-phenylamino benzenesulfonic acid sodium salt,diphenylaminesulfonic acid sodium salt PubChem CID: 517002 IUPAC Name: 4-(phenylamino)benzene-1-sulfonic acid sodium SMILES: [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Sodium diphenylamine-4-sulfonate, ACS Reagent, Solstice
CAS: 6152-67-6 Molecular Formula: C12H11NNaO3S Molecular Weight (g/mol): 272.27 MDL Number: MFCD00036468 InChI Key: VMVSDVIYIMNZBS-UHFFFAOYSA-N Synonym: sodium 4-phenylamino benzenesulfonate,sodium diphenylamine-4-sulfonate,benzenesulfonic acid, 4-phenylamino-, monosodium salt,unii-3e2ux1382o,sodium 4-anilinobenzenesulfonate,diphenylamine-4-sulfonic acid sodium salt,sodium diphenylamine-p-sulfonate,n-phenylsulfanilic acid, sodium salt,4-phenylamino benzenesulfonic acid sodium salt,diphenylaminesulfonic acid sodium salt PubChem CID: 517002 IUPAC Name: sodium;4-anilinobenzenesulfonate SMILES: [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1