Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate, MP Biomedicals

Catalog No. MP04811032 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04811032 500 g
MP04811033 25 g
MP04811030 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP04811032 Supplier MP Biomedicals, Inc Supplier No. 04811032
Add to Cart
Add to Cart

  • Used to solubilize and denature proteins for denaturing-PAGE
  • Most proteins bind SDS in a ratio of 1.4 g SDS per gram of protein
  • The charges intrinsic to the protein become insignificant compared to the overall negative charge provided by the bound SDS
  • The charge to mass ratio is essentially the same for each protein and will migrate in the gel based only on their size

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material SODIUM DODECYL SULFATE
Color White
Physical Form Powder
Melting Point 180°C to 210°C
Quantity 500 g
Molecular Formula C12H25NaO4S
Linear Formula C12H25O4SNa
MDL Number MFCD00036175
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
Solubility Information Soluble in water (200mg/mL - clear, faint yellow solution), and ethanol (0.1g/10 mL).
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
IUPAC Name sodium dodecyl sulfate
Molecular Weight (g/mol) 288.38
PubChem CID 3423265
ChEBI CHEBI:8984
Formula Weight 288.38
Percent Purity ≥99%
Grade Ultra Pure
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.