Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate, PlusOne™, Cytiva

Catalog No. 45000227 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-227 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-227 Supplier Cytiva Supplier No. 17131301
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 151-21-3 , 7732-18-5
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Quantity 100 g
Assay Percent Range ≥98.5% (sodium dodecyl sulfate C12 compounds)
Formula Weight 288.38
Chemical Name or Material Sodium Dodecyl Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.