Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™ PROMO

Catalog No. BP166100S24 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP166100S24 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP166100S24

Synonym: Sodium Lauryl Sulfate, Sodium Dodecylsulfate, Neutrazyme

  • DNase and RNase-free detergent for protein extraction procedures and polyacrylamide gel electrophoresis
  • SDS is a commonly used detergent in protein purification and polyacrylamide gel electrophoresis (PAGE)
  • SDS denatures proteins by disrupting the non-covalent bonds of the protein
  • Functionally tested for use in electrophoresis
  • Intended for research use only; not for human or animal diagnostic purposes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3 , 7647-14-5 , 7757-82-6
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium Lauryl Sulfate
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Absorbance 0.1 AU (0.1% Solution) at 280 nm
Color White
Physical Form Solid
Melting Point 206°C
pH 8.5 to 10
Quantity 100 g
Identification Pass Test
Packaging Glass Bottle
Linear Formula CH3(CH2)10CH2OSO3Na
Merck Index 15, 8768
Formula Weight 288.38
Percent Purity ≥99%
Grade DNase- and RNase-free
Assay ≥99% as Fatty Alcohol Sulfate
Chemical Name or Material Sodium Dodecyl Sulfate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement Flammable solid.
Precautionary Statement Causes eye irritation. Causes skin irritation. May cause respiratory irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.