Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Erythorbate, FCC, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606497 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-497 125 g Poly Bottle
18-606-498 500 g Poly Bottle
18-606-499 2.5 kg Poly Bottle
18-606-500 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-606-497 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1223125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

S1223, 6381-77-7, C6H7NaO6·H2O

Spectrum™ Chemical Sodium Erythorbate, FCC is used as an antioxidant preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6381-77-7
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
InChI Key QNSCGSPRKIPKTN-RKJRWTFHSA-N
IUPAC Name sodium (2R)-2-[(2R)-4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl]-2-hydroxyethan-1-olate
SMILES [Na+].O[C@H](C[O-])[C@H]1OC(O)=C(O)C1=O

Specifications

CAS Max % 1
Loss on Drying 0.0025
Packaging Poly Bottle
Quantity 125 g
Percent Purity 98 to 100.5%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sodium Erythorbate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.