Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium fluoride, Puratronic™, 99.995% (metals basis excluding K), K =100ppm, Thermo Scientific Chemicals

Catalog No. p-4407884
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1296406 5 g
AA1296418 50 g
AA1296430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1296406 Supplier Thermo Scientific Chemicals Supplier No. 01296406
Only null left
Add to Cart

It is used as a steel degreasing agent and a flux. It is used in heat treating salt compositions, tin-electroplating, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It has significant applications in organic synthesis including its halide-exchange with acyl chloride, sulfur chlorides, and phosphorus chloride, desilylation of silyl-protected OH groups, synthesis of fluorocarbons (Finkelstein reaction), and also in metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
SMILES [F-].[Na+]

Specifications

Boiling Point 1704°C
Melting Point 993°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9995% (metals basis excluding K)
Solubility Information Soluble in water. Slightly soluble in HF and ammonia. Insoluble in alcohol.
IUPAC Name sodium fluoride
Formula Weight 41.99
Odor Odorless
Refractive Index 1.336
Appearance White powder
Vapor Pressure 1.3 hPa (1mm Hg) at 20°C
Density 2.78
Chemical Name or Material Sodium fluoride
Show More Show Less
Hazard Category H301-H315-H319
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1, Packing Group: III
EINECSNumber 231-667-8
RTECSNumber WB0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.