Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Fluoride, Powder, USP, EP, BP, 98.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606506 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-506 45 kg Poly Bottle
18-606-503 500 g Poly Bottle
18-606-504 2.5 kg Poly Bottle
18-606-505 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-506 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO16745KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SO167, 7681-49-4, NaF

Spectrum™ Chemical Sodium Fluoride, Powder, USP, EP, BP is used to prevent cavities making teeth stronger and more resistant to decay caused by acid and bacteria. Toothpaste being the preferred delivery method. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 45 kg
Percent Purity 98.5 to 100.5%
Grade Multi-Compendial
Loss on Drying 0.005
Packaging Poly Bottle
Appearance Clear
Chemical Name or Material Sodium Fluoride, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.