Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Formate, ≥99%, ACS Reagent, Solstice

Catalog No. 60045655 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-655 100 g Poly Bottle
60-045-657 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-655 Supplier Solstice Supplier No. 247596100G

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.007
MDL Number MFCD00013101
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
PubChem CID 2723810
ChEBI CHEBI:62965
IUPAC Name sodium;formate
SMILES C(=O)[O-].[Na+]

Specifications

Melting Point 259°C to 262°C
Color White
Quantity 100 g
Linear Formula HCOONa
UN Number NONH for all modes of transport
Beilstein 3595134
Solubility Information Soluble in water
Formula Weight 68.01g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder or crystals
Density 1.92g/cm3
Chemical Name or Material Sodium Formate
Show More Show Less
EINECSNumber 205-488-0
RTECSNumber LR0350000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.